Merck
  • AP-1 pathway-targeted inhibition of inflammatory responses in LPS-treated macrophages and EtOH/HCl-treated stomach by Archidendron clypearia methanol extract.
AP-1 pathway-targeted inhibition of inflammatory responses in LPS-treated macrophages and EtOH/HCl-treated stomach by Archidendron clypearia methanol extract.
Journal of ethnopharmacology (2013-02-16)
Woo Seok Yang, Deok Jeong, Gyeongsug Nam, Young-Su Yi, Deok Hyo Yoon, Tae Woong Kim, Yung Chul Park, Hyunsik Hwang, Man Hee Rhee, Sungyoul Hong, Jae Youl Cho
ABSTRACT
Archidendron clypearia Jack. (Fabaceae) is a representative ethnomedicinal herbal plant prescribed for various inflammatory diseases such as pharyngolaryngitis and tonsillitis. However, the pharmacology behind this plant's anti-inflammatory properties has not been fully understood. Therefore, in this study, the anti-inflammatory mechanism of a 95% methanol extract (Ac-ME) was explored. The anti-inflammatory mechanism of Ac-ME on the AP-1 activation pathway, which plays a critical role in the production of prostaglandin (PG)E2 in RAW264.7 cells and peritoneal macrophages and in induction of acute gastritis caused by HCl/EtOH, was investigated using immunoblotting, immunoprecipitation analyses, and reporter gene activity assays. In particular, enzyme assays and HPLC analysis were employed to identify direct target enzymes of Ac-ME and to detect active chemical components from the plant extract. Ac-ME clearly reduced the nuclear levels of total and phospho-forms of c-Jun, FRA-1, and ATF-2. Consequently, this extract suppressed both the production of PGE2 in lipopolysaccharide (LPS)-activated RAW264.7 and peritoneal macrophage cells and PGE2-dependent induction of gastritis lesion in stomach under EtOH/HCl exposure. Analysis of AP-1 upstream signalling revealed that the AP-1 activation pathway consisting of IRAK1, TRAF6, TAK1, MKK3/6, and p38 was predominantly inhibited by Ac-ME. Similarly, this extract directly blocked the enzyme activity of IRAK1, indicating that this enzyme is an inhibitory target of Ac-ME and is involved in the suppression of the AP-1 pathway. HPLC analysis showed that quercetin, which inhibits PGE2 production, is an active component in Ac-ME. Ac-ME is an ethnomedicinal remedy with an IRAK1/p38/AP-1-targeted inhibitory property. Since AP-1 is a major inflammation-inducing transcription factor, the therapeutic potential of Ac-ME in other AP-1-mediated inflammatory symptoms will be further tested.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, for GC derivatization, LiChropur