Merck
  • Content of polychlorinated dibenzo-p-dioxins, dibenzofurans and dioxin-like polychlorinated biphenyls in fish from Latvian lakes.
Content of polychlorinated dibenzo-p-dioxins, dibenzofurans and dioxin-like polychlorinated biphenyls in fish from Latvian lakes.
Chemosphere (2013-01-23)
D Zacs, V Bartkevics, A Viksna
ABSTRACT
Seventeen polychlorinated dibenzo-p-dioxins and polychlorinated dibenzofurans (PCDD/PCDFs) of the highest priority as well as twelve dioxin-like polychlorinated biphenyls (dl-PCBs) were analyzed in the muscle tissues of the following freshwater fish species sampled from eleven Latvian freshwater lakes: perch (Perca flavescens), carp (Cyprinus carpio), eel (Anguilla rostrata), bream (Abramis brama), chub (Leuciscus cephalus), pike (Esox lucius), sheatfish (Silurus glanis) and roach (Rutilus). To analyze the selected persistent organic pollutants in fish matrices, an optimization of EPA-1613 and EPA-1668A clean-up procedures was carried out, followed by validation of the analytical procedure according to Commission Regulation (EC) No 1883/2006. The adopted analytical procedure was in compliance with requirements of the more recent Commission Regulation (EU) No 252/2012. Modifications of carbon column chromatography clean-up and separation steps were used for treatment of the fish samples. Other clean-up procedure stages were performed according to the methods EPA-1613 and EPA-1668A and involved gel permeation chromatography (GPC), as well as manual acidic silica and Florisil column chromatography for purification and fractionation of the samples. An isotope dilution method was used for the qualitative and quantitative determination of individual congeners. Analytes of interest were separated and detected using gas chromatography - high resolution mass spectrometry. The concentration of PCDD/PCDFs and dl-PCBs in freshwater fish and eel samples ranged from 0.05 to 8.0 pg WHO(1998)-PCDD/PCDF-PCB-TEQ g(-1) fresh weight. These levels are below the EU maximum permissible limits although calculation of the content of these compounds relative to the weight of fat shows contamination levels similar to those found in Baltic herring and sprats, that are known to be highly contaminated. A difference in congener pattern between the Baltic Sea fish and freshwater fish was detected with lower contribution of 2,3,4,7,8-PeCDF to the contamination of inland water fish.
Product Description
Dibenzo[b,d]furan, BCR®, certified reference material
Sigma-Aldrich
Dibenzofuran, 98%