Merck
  • Valproic acid reduces the tolerability of temsirolimus in children and adolescents with solid tumors.
Valproic acid reduces the tolerability of temsirolimus in children and adolescents with solid tumors.
Anti-cancer drugs (2013-01-19)
Don W Coulter, Christine Walko, Jai Patel, Billie M Moats-Staats, Andrew McFadden, Scott V Smith, Wasiuddin A Khan, Arlene S Bridges, Allison M Deal, Javier Oesterheld, Ian J Davis, Julie Blatt
ABSTRACT
A pediatric study has established a maximum tolerated dose (MTD) for temsirolimus (Tem) of more than 150 mg/m intravenously/week. A phase I trial was conducted to establish the MTD for Tem in combination with valproic acid (VPA) in children and adolescents with refractory solid tumors. The secondary aims included expression of mammalian target of rapamycin (mTOR) markers on archival tumor tissue; Tem pharmacokinetics; assessment of histone acetylation (HA); and tumor response. Patients were treated with VPA (5 mg/kg orally three times daily) with a target serum level of 75-100 mcg/ml. Tem was started at an initial dose of 60 mg/m/week. Pharmacokinetics and HA measurements were performed during weeks 1 and 5. Two of the first three patients experienced dose-limiting toxicity (grade 3 mucositis). Tem at 35 mg/m/week was found to be tolerable. Peak Tem concentrations were higher in all patients compared with those in previously published reports of single agent Tem. Increases in HA are correlated with VPA levels. All tumor samples expressed mTORC1 and mTORC2. An objective response was observed in one patient (melanoma), whereas transient stable disease was observed in four other patients (spinal cord ependymoma, alveolar soft part sarcoma, medullary thyroid carcinoma, and hepatocellular carcinoma). The MTD of Tem when administered with VPA is considerably lower than when used as a single agent, with mucositis the major dose-limiting toxicity. The combination merits further study and may have activity in melanoma. Attention to drug-drug interactions will be important in future multiagent trials including Tem.
Product Description
Supelco
Diphenhydramine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.
Sigma-Aldrich
Diphenhydramine hydrochloride, ≥98% (HPLC)
Pricing and availability is not currently available.
Supelco
Diphenhydramine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Diphenhydramine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.