Merck
  • Pseudolarenone, an unusual nortriterpenoid lactone with a fused 5/11/5/6/5 ring system featuring an unprecedented bicyclo[8.2.1]tridecane core from Pseudolarix amabilis.
Pseudolarenone, an unusual nortriterpenoid lactone with a fused 5/11/5/6/5 ring system featuring an unprecedented bicyclo[8.2.1]tridecane core from Pseudolarix amabilis.
Chemical communications (Cambridge, England) (2013-01-04)
Bo Li, De-Yun Kong, Yun-Heng Shen, Ka-Li Fu, Rong-Cai Yue, Zhu-Zhen Han, Hu Yuan, Qing-Xin Liu, Lei Shan, Hui-Liang Li, Xian-Wen Yang, Wei-Dong Zhang
ABSTRACT
A phytochemical investigation of the cone of Pseudolarix amabilis led to the isolation of pseudolarenone (), a structurally novel pentacyclic (5/11/5/6/5) nortriterpenoid lactone with an unprecedented carbon skeleton featuring a unique bicyclo[8.2.1]tridecane core. Its structure and absolute configurations were elucidated by spectroscopic analysis and single crystal X-ray diffraction (CuK(α)).
Product Description
Supelco
Tridecane, analytical standard
Sigma-Aldrich
Tridecane, ≥99%