Merck
  • [Recent researches of synthetic mercury sulfide in traditional medicine system].
[Recent researches of synthetic mercury sulfide in traditional medicine system].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2012-12-29)
Chao-jun Chen, Shi-kui Wu, Yi-bo Wang, Jin-feng Hou, Lei Ma, Xiao-yan Sun
ABSTRACT
Herein, the synthesis, component, microstructure and pharmacological and toxicology researches of the Synthetic Mercury Sulfide (S-HgS) a kind of common drug in Chinese, Mongolia, Tibetan medicine, and Indian medicine system were summarized. The similar cognition about mercury toxicity & pharmacological action from some Asian regions was analyzed, and it can supply some useful direction for the traditional Asian medicine system. Recent literatures both domestic and abroad were summarized and analyzed. S-HgS is the basis of Vermilion, Mongolia-Vermilion, Zuotai, and Ras-sindoor. Athough the processes of synthesis are very different, but the microstructure and pharmacological & toxicology of S-HgS is similar. S-HgS has a far-ranging application,and unique curative effect. New technology such as nanotechnology can be used for improving the advancement of traditional Asian medicine.
Sigma-Aldrich
Mercury(II) sulfate, puriss., ≥98%
Supelco
Mercury(II) sulfate, ACS reagent, for preparation of solution for COD determination according to DIN 38409, part 41, ≥99%