Merck
  • Consensus on the use of the fixed combination calcipotriol/betamethasone dipropionate in the treatment of plaque psoriasis.
Consensus on the use of the fixed combination calcipotriol/betamethasone dipropionate in the treatment of plaque psoriasis.
Giornale italiano di dermatologia e venereologia : organo ufficiale, Societa italiana di dermatologia e sifilografia (2012-11-15)
G Girolomoni, G A Vena, F Ayala, S P Cannavò, O De Pità, S Chimenti, A Peserico
ABSTRACT
Calcipotriol, a vitamin D analogue, and betamethasone dipropionate, a high potency corticosteroid, are complementary agents for the topical treatment of psoriasis vulgaris. Robust evidence on the efficacy and safety of their fixed combination has been provided by randomized, double-blind, controlled clinical trials involving more than 7000 patients with the ointment formulation in psoriasis of the body and more than 4000 patients with the gel formulation in scalp psoriasis. These trials have shown that the fixed combination ointment is more effective and better tolerated, not only than placebo, but also than calcipotriol and tacalcitol monotherapies. In addition, it has proved, in most instances, to be more effective than betamethasone and at least as well tolerated. The same applies to the gel for scalp and body psoriasis. Safety studies have excluded that repeated courses of treatment with the fixed combination for up to one year produce systemic effects. Studies have also shown that the fixed combination treatment improves quality of life to a significantly greater extent than calcipotriol, with the once daily regimen most appreciated by patients, in both active disease and recurrency. Because of the extensive evidence, American and European guidelines recommend the calcipotriol/betamethasone dipropionate fixed combination as first line topical treatment for mild to moderate plaque psoriasis of the body and scalp.
Sigma-Aldrich
Calcipotriol hydrate, ≥98% (HPLC)
Betamethasone dipropionate for peak identification, European Pharmacopoeia (EP) Reference Standard
USP
Betamethasone dipropionate, United States Pharmacopeia (USP) Reference Standard
Supelco
Betamethasone, VETRANAL®, analytical standard
Betamethasone dipropionate, European Pharmacopoeia (EP) Reference Standard
Betamethasone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Betamethasone, ≥98%
Sigma-Aldrich
Betamethasone 17,21-dipropionate
Supelco
Betamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Betamethasone Dipropionate, Pharmaceutical Secondary Standard; Certified Reference Material