Merck
  • Tetraphenylethene modified perylene bisimide: effect of the number of substituents on AIE performance.
Tetraphenylethene modified perylene bisimide: effect of the number of substituents on AIE performance.
Chemical communications (Cambridge, England) (2012-10-26)
Qiuli Zhao, Xiao A Zhang, Qiang Wei, Jian Wang, Xiao Yuan Shen, Anjun Qin, Jing Zhi Sun, Ben Zhong Tang
ABSTRACT
Perylene bisimide derivatives substituted with one and two tetraphenylethene moieties at 1 and 1,7-postions show distinct optical properties. The former displays characteristic emission features of perylene bisimides in solution and red emission in the aggregate state, while the latter is nonemissive in solution but highly red-emissive in the aggregate state.
Sigma-Aldrich
Anthracene, sublimed grade, ≥99%
Supelco
Anthracene, analytical standard
Supelco
Anthracene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Anthracene, reagent grade, 97%
Sigma-Aldrich
Anthracene, ReagentPlus®, 99%
Supelco
Anthracene, analytical standard
Sigma-Aldrich
Anthracene, suitable for scintillation, ≥99.0% (GC)