Merck
  • Feasibility of carbidopa premedication in pediatric patients: a pilot study.
Feasibility of carbidopa premedication in pediatric patients: a pilot study.
Cancer biotherapy & radiopharmaceuticals (2012-10-06)
Egesta Lopci, Daniela D'Ambrosio, Cristina Nanni, Arturo Chiti, Andrea Pession, Mario Marengo, Stefano Fanti
ABSTRACT
To verify the potential role and feasibility of carbidopa premedication in pediatric patients undergoing ¹⁸F-DOPA (Fluorine-18 fluorodihydroxyphenylalanine) PET scanning. For this limited study, 5 patients (M:F=3:2; mean age 4.8 years) with a positive history for neuroblastoma who had been referred to our institution for instrumental monitoring during clinical follow-up were enrolled. In all cases, two consecutive ¹⁸F-DOPA PET scans, the first without carbidopa and the second with carbidopa premedication, were scheduled: patients received 4 MBq/kg of radiotracer and a dose of 2 mg/kg of carbidopa. Dedicated VOIs were drawn on the basal ganglia, pancreas, liver, and renal cortex. These regions were semiquantitatively analyzed at both the first and at the second ¹⁸F-DOPA scan, and mean SUV(max) values were compared using the t-test. On a visual basis, a clear reduction in the abdominal accumulation of (18)F-DOPA was observed in all cases after carbidopa premedication. This reduction related both to the biliary structures and the excretory system, and was accompanied by a generalized increase in soft tissue uptake. The semiquantitative analysis documented an absolute increase in SUV(max) after carbidopa premedication in the basal ganglia (3.4±1.3 vs. 2.1±0.8) and liver parenchyma (2.2±0.5 vs. 1.5±0.5), whereas SUV(max) decreased in the renal cortex (1.7±0.8 vs. 3.7±1.0) and the pancreas (2.3±0.6 vs. 3.5±0.5). The changes in SUV(max) were statistically significant for the pancreas and liver parenchyma (p=0.022 and 0.045, respectively), but not for the basal ganglia and renal cortex (p=0.143 and 0.15, respectively). Carbidopa premedication in the pediatric population appears feasible and seems to influence ¹⁸F-DOPA distribution in the liver and pancreas in a manner similar to that reported in adults. Larger series are however needed to properly define the clinical role of carbidopa premedication in children.
Product Description
Carbidopa, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
S-(−)-Carbidopa, ≥98% (TLC), powder