Merck
  • Conversion and kinetics study of fructose-to-5-hydroxymethylfurfural (HMF) using sulfonic and ionic liquid groups bi-functionalized mesoporous silica nanoparticles as recyclable solid catalysts in DMSO systems.
Conversion and kinetics study of fructose-to-5-hydroxymethylfurfural (HMF) using sulfonic and ionic liquid groups bi-functionalized mesoporous silica nanoparticles as recyclable solid catalysts in DMSO systems.
Physical chemistry chemical physics : PCCP (2012-09-20)
Yin-Ying Lee, Kevin C-W Wu
ABSTRACT
Mesoporous silica nanoparticles functionalized with both sulfonic acid (HSO(3)) and ionic liquid (ILs) were synthesized and applied as effective and recyclable catalysts for generating 5-hydroxymethylfurfural (HMF) from fructose. For the first time a high HMF yield of 72.5% was achieved in DMSO systems under mild conditions (90 °C and 3 h). We further studied the kinetics of the fructose-to-HMF conversion and compared the rate constants, reaction orders, and activation energies for the systems with and without bi-functional MSN catalysts.
Sigma-Aldrich
5-(Hydroxymethyl)furfural, ≥99%, FG
Supelco
5-(Hydroxymethyl)furfural, analytical standard
Sigma-Aldrich
5-Hydroxymethyl-2-furaldehyde, 99%