Merck
  • Cross-coupling of diarylborinic acids and anhydrides with arylhalides catalyzed by a phosphite/N-heterocyclic carbene co-supported palladium catalyst system.
Cross-coupling of diarylborinic acids and anhydrides with arylhalides catalyzed by a phosphite/N-heterocyclic carbene co-supported palladium catalyst system.
The Journal of organic chemistry (2012-08-07)
Xiaofeng Chen, Haihua Ke, Yao Chen, Changwei Guan, Gang Zou
ABSTRACT
A highly efficient cross-coupling of diarylborinic acids and anhydrides with aryl chlorides and bromides has been effected by using a palladium catalyst system co-supported by a strong σ-donor N-heterocyclic carbene (NHC), N,N'-bis(2,6-diisopropylphenyl) imidazol-2-ylidene, and a strong π-acceptor phosphite, triphenylphosphite, in tert-BuOH in the present of K(3)PO(4)·3H(2)O. Unsymmetrical biaryls with a variety of functional groups could be obtained in good to excellent yields using as low as 0.01, 0.2-0.5, and 1 mol % palladium loadings for aryl bromides and activated and deactivated aryl chlorides, respectively, under mild conditions. A ligand synergy between the σ-donor NHC and the π-acceptor phosphite in the Pd/NHC/P(OPh)(3) catalytic system has been proposed to be responsible for the high efficacy to arylchlorides in the cross-coupling. A scalable and economical process has therefore been developed for synthesis of Sartan biphenyl from the Pd/NHC/P(OPh)(3) catalyzed cross-coupling of di(4-methylphenyl)borinic acid with 2-chlorobenzonitrile.
Sigma-Aldrich
Biphenyl, ≥99%
Supelco
Biphenyl, PESTANAL®, analytical standard
Sigma-Aldrich
Biphenyl, ReagentPlus®, 99.5%