Merck
  • Synthesis, design and biological evaluation of novel highly potent tacrine congeners for the treatment of Alzheimer's disease.
Synthesis, design and biological evaluation of novel highly potent tacrine congeners for the treatment of Alzheimer's disease.
European journal of medicinal chemistry (2012-07-24)
Slavka Hamulakova, Ladislav Janovec, Martina Hrabinova, Pavol Kristian, Kamil Kuca, Maria Banasova, Jan Imrich
ABSTRACT
New tacrine derivatives 5a-d, 6a-d with piperazino-ethyl spacer linked with corresponding secondary amines and tacrine homodimer 8 were synthesized and tested as cholinesterase inhibitors on human acetylcholinesterase (hAChE) and human plasmatic butyrylcholinesterase (hBChE). In most cases the majority of synthesized derivatives exhibit a high AChE and BChE inhibitory activity with IC(50) values in the low-nanomolar range, being clearly more potent than the reference standard tacrine (9-amino-1,2,3,4-tetrahydroacridine, 1) and 7-MEOTA (7-methoxy-9-amino-1,2,3,4-tetrahydroacridine). Among them, inhibitors 8 and 5c, showed a strong inhibitory activity against hAChE, with an IC(50) value of 4.49 nM and 4.97, nM resp., and a high selectivity to hAChE. The compound 5d acted as the most potent inhibitor against hBChE with an IC(50) value of 33.7 nM and exhibited also a good selectivity towards hBChE. The dissociation constants K(i) of the selected inhibitors were compared with their IC(50) values. Molecular modeling studies were performed to predict the binding modes between individual derivatives and hAChE/hBChE.
Product Description
Supelco
Piperazine hexahydrate, analytical standard
Sigma-Aldrich
Piperazine, ReagentPlus®, 99%
Sigma-Aldrich
Piperazine, BioUltra, anhydrous, ≥99.0% (T)