Merck
  • BODIPY based colorimetric fluorescent probe for selective thiophenol detection: theoretical and experimental studies.
BODIPY based colorimetric fluorescent probe for selective thiophenol detection: theoretical and experimental studies.
The Analyst (2012-07-04)
Dnyaneshwar Kand, Pratyush Kumar Mishra, Tanmoy Saha, Mayurika Lahiri, Pinaki Talukdar
ABSTRACT
A BODIPY-based selective thiophenol probe capable of discriminating aliphatic thiols is reported. The fluorescence off-on effect upon reaction with thiol is elucidated with theoretical calculations. The sensing of thiophenol is associated with a color change from red to yellow and 63-fold enhancement in green fluorescence. Application of the probe for selective thiophenol detection is demonstrated by live cell imaging.
Sigma-Aldrich
Sodium thiophenolate, technical grade, 90%
Sigma-Aldrich
Thiophenol, 97%
Supelco
Thiophenol, analytical standard
Sigma-Aldrich
Thiophenol, ≥99%