Merck
  • Clioquinol induces DNA double-strand breaks, activation of ATM, and subsequent activation of p53 signaling.
Clioquinol induces DNA double-strand breaks, activation of ATM, and subsequent activation of p53 signaling.
Toxicology (2012-05-26)
Masato Katsuyama, Kazumi Iwata, Masakazu Ibi, Kuniharu Matsuno, Misaki Matsumoto, Chihiro Yabe-Nishimura
ABSTRACT
Clioquinol, a Cu²⁺/Zn²⁺/Fe²⁺ chelator/ionophor, was used extensively in the mid 1900s as an amebicide for treating indigestion and diarrhea. It was eventually withdrawn from the market because of a link to subacute myelo-optic neuropathy (SMON) in Japan. The pathogenesis of SMON, however, is not fully understood. To clarify the molecular mechanisms of clioquinol-induced neurotoxicity, a global analysis using DNA chips was carried out on human neuroblastoma cells. The global analysis and quantitative PCR demonstrated that mRNA levels of p21(Cip1), an inhibitor of cyclins D and E, and of GADD45α, a growth arrest and DNA damage-inducible protein, were significantly increased by clioquinol treatment in SH-SY5Y and IMR-32 neuroblastoma cells. Activation of p53 by clioquinol was suggested, since clioquinol induced phosphorylation of p53 at Ser15 to enhance its stabilization. The phosphorylation of p53 was inhibited by KU-55933, an inhibitor of ataxia-telangiectasia mutated kinase (ATM), but not by NU7026, an inhibitor of DNA-dependent protein kinase (DNA-PK). Clioquinol in fact induced phosphorylation of ATM and histone H2AX, a marker of DNA double-strand breaks (DSBs). These results suggest that clioquinol-induced neurotoxicity is mediated by DSBs and subsequent activation of ATM/p53 signaling.
Product Description
Supelco
Clioquinol, VETRANAL®, analytical standard
Clioquinol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
5-Chloro-7-iodo-8-quinolinol, ≥95.0% (HPLC)