Merck
  • Efficient capture of SO2 by a binary mixture of caprolactam tetrabutyl ammonium bromide ionic liquid and water.
Efficient capture of SO2 by a binary mixture of caprolactam tetrabutyl ammonium bromide ionic liquid and water.
Journal of hazardous materials (2011-08-27)
Erhong Duan, Bin Guo, Miaomiao Zhang, Yanan Guan, Hua Sun, Jing Han
ABSTRACT
The solubility of SO(2) in a binary mixture of water and caprolactam tetrabutyl ammonium bromide ionic liquid (CPL-TBAB IL) was investigated. Though the ionic liquid and water were fully miscible, a phase separation occurred when SO(2) was introduced into the mixture. The SO(2) concentrated in the lower layer, and it could be released by heating the solution under reduced pressure (382.2K, 10.1 kPa). After desorption, the mixture could be reused to absorb SO(2). It was found that SO(2) acts as a switch to cause the water and CPL-TBAB IL to phase separate, and the mechanics of this phase separation process was studied by gas chromatography-mass spectrometry, fourier transform-infrared spectroscopy and Karl-Fisher titration. The absorption and desorption of SO(2) in the CPL-TBAB/water mixtures were reversible.
Sigma-Aldrich
Ammonium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium bromide, 99.999% trace metals basis
Sigma-Aldrich
Ammonium bromide, ≥99.99% trace metals basis