Merck
  • Iodobismuthates with N-alkyl- or N,N'-dialkyl-4,4'-bipyridinium: syntheses, structures and dielectric properties.
Iodobismuthates with N-alkyl- or N,N'-dialkyl-4,4'-bipyridinium: syntheses, structures and dielectric properties.
Physical chemistry chemical physics : PCCP (2011-02-12)
Yang Chen, Zhou Yang, Xin-Yi Wu, Chun-Yan Ni, Zhi-Gang Ren, Hui-Fang Wang, Jian-Ping Lang
ABSTRACT
The solvothermal reactions of BiI(3), KI, I(2), 4,4'-bipyridine (4,4'-bipy), and a small amount of water in alcohol and acetonitrile produced four bipyridinium iodobismuthates {[MQ](3)[Bi(2)I(6)(μ-I)(3)][Bi(2)I(6)(μ-I)(2)(MQ)(2)](3)} (1, MQ(+) = N-methyl-4,4'-bipyridinium), {[EQ](3)[Bi(2)I(6)(μ-I)(3)][Bi(2)I(6)(μ-I)(2)(EQ)(2)](3)} (2, EQ(+) = N-ethyl-4,4'-bipyridinium), [MV][BiI(5)] (Eur. J. Inorg. Chem., 2010, 5326) (3, MV(2+) = N,N'-dimethyl-4,4'-bipyridinium), and [EV](2)[Bi(4)I(10)(μ-I)(4)(μ(3)-I)(2)] (4, EV(2+) = N,N'-diethyl-4,4'-bipyridinium). In these reactions, 4,4'-bipy was partly or completely alkylated by alkyl groups generated from the cleavage of C-O bond of alcohols, forming the N-alkyl-4,4'-bipyridinium cation (Q(+)) and the N,N'-dialkyl-4,4'-bipyridinium dication (V(2+)), respectively. Compounds 1-4 were characterized by elemental analysis, IR, (1)H NMR and single-crystal X-ray diffraction analysis. The optical, electrical conductive and dielectric properties of these compounds were investigated. The dielectric constants of the Q(+)-based compounds were larger than the values of the V(2+)-based ones, which showed that the weak electrostatic interactions in the structures may benefit the polarizability of molecules, thereby resulting in a larger dielectric response of the structures under an external electric field, while the strong electrostatic interactions between the positive and negative charge units would lead to a low dielectric constant (low-k) behavior of these compounds.
Sigma-Aldrich
Bismuth(III) iodide, ≥99.998% trace metals basis
Sigma-Aldrich
Bismuth(III) iodide, 99%