Merck
  • Poly(D,L-lactide-co-glycolide)/hydroxyapatite core-shell nanosphere. Part 2: Simultaneous release of a drug and a prodrug (clindamycin and clindamycin phosphate).
Poly(D,L-lactide-co-glycolide)/hydroxyapatite core-shell nanosphere. Part 2: Simultaneous release of a drug and a prodrug (clindamycin and clindamycin phosphate).
Colloids and surfaces. B, Biointerfaces (2010-10-19)
Marija Vukomanović, Srečo Davor Skapin, Ida Poljanšek, Ema Zagar, Bogdan Kralj, Nened Ignjatović, Dragan Uskoković
ABSTRACT
The novel concept of a simultaneous, controlled release of a drug and a prodrug with different physico-chemical properties was applied in order to prolong the release period of antibiotics and estimate their high local concentrations, which are the necessary preconditions for the treatment of some chronic infection diseases. For this purpose poly(D,L-lactide-co-glycolide)/hydroxyapatite (PLGA/HAp) core-shell nanostructures were used as the carrier of clindamycin-base, as a drug, and clindamycin-2-phosphate, as a prodrug model. As a result, a two-step release was observed: the controlled release of the more soluble phosphate form and the sustained release of the less-soluble base form of clindamycin, resulting in a high overall concentration of the released drug during the period of 30 days in vitro. The HAp phase within the PLGA core-shells, applied as a drug carrier, delayed the process of the degradation of the polymer; however, the presence of the drug affected the process of degradation and this influence was the dominant factor in the control over the degradation of the polymer phase of PLGA/HAp and the consequent kinetics of the drug release.
Sigma-Aldrich
Clindamycin 2-phosphate, aminoglycoside antibiotic
Clindamycin phosphate, European Pharmacopoeia (EP) Reference Standard
Supelco
Clindamycin Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material