Merck
  • Regioselective allene synthesis and propargylations with propargyl diethanolamine boronates.
Regioselective allene synthesis and propargylations with propargyl diethanolamine boronates.
Organic letters (2009-11-03)
Daniel R Fandrick, Jonathan T Reeves, Zhulin Tan, Heewon Lee, Jinhua J Song, Nathan K Yee, Chris H Senanayake
ABSTRACT
The utility of propargyl diethanolamine boronates as reagents for the preparation of allenes and homopropargylic alcohols is presented. Protonolysis with TFA and electrophilic substitution with N-halosuccinimides proceeded with inversion to provide the corresponding allene in high yield and regioselectivity. Alternatively, the propargylation of aldehydes was achieved with use of the in situ generated lithiated complex.
Product Description
Supelco
Diethanolamine, analytical standard
Sigma-Aldrich
Diethanolamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Diethanolamine, reagent grade, ≥98.0%
Sigma-Aldrich
Diethanolamine, ACS reagent, ≥98.5%
Sigma-Aldrich
Diethanolamine, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Diethanolamine hydrochloride, 98%