Merck
  • Ceramide starves cells to death by downregulating nutrient transporter proteins.
Ceramide starves cells to death by downregulating nutrient transporter proteins.
Proceedings of the National Academy of Sciences of the United States of America (2008-11-05)
Garret G Guenther, Eigen R Peralta, Kimberly Romero Rosales, Susan Y Wong, Leah J Siskind, Aimee L Edinger
ABSTRACT
Ceramide induces cell death in response to many stimuli. Its mechanism of action, however, is not completely understood. Ceramide induces autophagy in mammalian cells maintained in rich media and nutrient permease downregulation in yeast. These observations suggested to us that ceramide might kill mammalian cells by limiting cellular access to extracellular nutrients. Consistent with this proposal, physiologically relevant concentrations of ceramide produced a profound and specific downregulation of nutrient transporter proteins in mammalian cells. Blocking ceramide-induced nutrient transporter loss or supplementation with the cell-permeable nutrient, methyl pyruvate, reversed ceramide-dependent toxicity. Conversely, cells became more sensitive to ceramide when nutrient stress was increased by acutely limiting extracellular nutrients, inhibiting autophagy, or deleting AMP-activated protein kinase (AMPK). Observations that ceramide can trigger either apoptosis or caspase-independent cell death may be explained by this model. We found that methyl pyruvate (MP) also protected cells from ceramide-induced, nonapoptotic death consistent with the idea that severe bioenergetic stress was responsible. Taken together, these studies suggest that the cellular metabolic state is an important arbiter of the cellular response to ceramide. In fact, increasing nutrient demand by incubating cells in high levels of growth factor sensitized cells to ceramide. On the other hand, gradually adapting cells to tolerate low levels of extracellular nutrients completely blocked ceramide-induced death. In sum, these results support a model where ceramide kills cells by inducing intracellular nutrient limitation subsequent to nutrient transporter downregulation.
Sigma-Aldrich
Methyl pyruvate, 90%, technical grade