Merck
LC-ESI-MS determination and pharmacokinetics of adrafinil in rats.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2008-08-30)
R Nageswara Rao, Dhananjay D Shinde, M V N Kumar Talluri, Sachin B Agawane
ABSTRACT
A highly sensitive and specific liquid chromatography/tandem mass spectrometric (LC-MS/MS) method for investigating the pharmacokinetics of adrafinil in rats was developed. Rat serum pretreated by solid-phase extraction (SPE) was analyzed by LC-MS/MS with an electrospray ionization (ESI) interface. The mobile phase consisted of acetonitrile:water:acetic acid (35:65:0.1, v/v/v) in an isocratic elution mode pumped at 1.0 ml/min. The analytical column (250 mm x 4.6 mm i.d.) was packed with Kromasil C(18) material (5.0 microm). The standard curve was linear from 16.5 to 5000 ng/ml. The assay was specific, accurate (R.S.D.<2.6%), precise and reproducible (within- and between-day precisions R.S.D. <7.0% and <9.0%, respectively). Adrafinil in rat serum was stable over three freeze-thaw cycles at ambient temperature for 6 h. The method had a lower limit of quantitation of 16.5 ng/ml, which offered high sensitivity for the determination of adrafinil in serum. The method was successfully applied to pharmacokinetic studies of adrafinil after an oral administration to rats.
Product Description
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 2.1 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4.6 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 3 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4.6 mm
Supelco
Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 21.2 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 5 cm × 2.1 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 2.1 mm
Supelco
Discovery® C18 Supelguard Guard Cartridge, Kit, 5 μm particle size, L × I.D. 2 cm × 3 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 10 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 12.5 cm × 4 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 10 cm × 4 mm
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea
Supelco
Discovery® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm, pkg of 2 ea
Supelco
Discovery® C18 Supelguard Guard Cartridge, 5 μm particle size, L × I.D. 2 cm × 3 mm, pkg of 2 ea
Supelco
Discovery® C18 HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm