Merck
  • Isolation, structure elucidation, and synthesis of cytotoxic tryptanthrin analogues from Phaius mishmensis.
Isolation, structure elucidation, and synthesis of cytotoxic tryptanthrin analogues from Phaius mishmensis.
Journal of natural products (2008-05-30)
Chen-Wei Jao, Wei-Chih Lin, Yao-Ting Wu, Pei-Lin Wu
ABSTRACT
Bioassay-guided chromatographic separation of the cytotoxic MeOH extract of Phaius mishmensis led to the isolation of two known and six new indoloquinazolinones, phaitanthrins A-E (1-5) and methylisatoid (6). The structures of the new compounds were elucidated by spectroscopic analysis. Phaitanthrin A (1) and tryptanthrin (7) showed moderate cytotoxicity against MCF-7, NCI-H460, and SF-268 cell lines. A series of ketone adducts of tryptanthrin were prepared and tested initially for anticancer activity in vitro against MCF-7, NCI-H460, and SF-268 human cancer cell lines. The 3-pentanone adduct 13 showed activity similar to tryptanthrin.
Sigma-Aldrich
Tryptanthrin, ≥98% (HPLC)