Merck
Phenolic and other constituents of fresh water fern Salvinia molesta.
Phytochemistry (2008-01-08)
M Iqbal Choudhary, Nadra Naheed, Ahmed Abbaskhan, Syed Ghulam Musharraf, Hina Siddiqui, Atta-Ur-Rahman
ABSTRACT
Two glycosides, 6'-O-(3,4-dihydroxy benzoyl)-beta-D-glucopyranosyl ester (1), and 4-O-beta-d-glucopyranoside-3-hydroxy methyl benzoate (2), along with five known compounds methyl benzoate (3), hypogallic acid (4), caffeic acid (5), paeoniflorin (6) and pikuroside (7) were isolated for the first time from a fresh water fern Salvinia molesta D.S. Mitch. These compounds showed a potent antioxidant radical scavenging activity in a non-physiological assay. Their structures were determined by NMR spectroscopic and CID mass spectrometry techniques.
Sigma-Aldrich
Methyl benzoate, 99%
Sigma-Aldrich
Methyl benzoate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Methyl benzoate, ≥98%, FCC, FG
Supelco
Methyl benzoate, analytical standard