Merck
  • Sensitive, quantitative, and fast modifications for Coomassie Blue staining of polyacrylamide gels.
Sensitive, quantitative, and fast modifications for Coomassie Blue staining of polyacrylamide gels.
Proteomics (2006-10-13)
Reiner Westermeier
ABSTRACT
In spite of the high sensitivity of silver staining and the wide dynamic range of various fluorescent detection methods, Coomassie Brilliant Blue staining is still the most widely used protein detection technique for proteins separated by polyacrylamide gel electrophoresis. There are several reasons: Low price, Visible with the eye, Desk top scanners can be employed for image acquisition, Better for quantitative analysis than silver staining, Possible modifications for fast or highly sensitive staining, Mass spectrometry compatible.
Sigma-Aldrich
Brilliant Blue R Staining Solution, suitable for (for immunoelectrophoresis protein staining)
Sigma-Aldrich
Brilliant Blue R, Dye content ~50 %, Technical grade
Sigma-Aldrich
Brilliant Blue R, pure
Sigma-Aldrich
Brilliant Blue R, 250, for microscopy
Sigma-Aldrich
Brilliant Blue R Concentrate, suitable for SDS-PAGE, methanol solution