Merck
Laterality of the olfactory event-related potential response.
Chemical senses (2006-07-20)
Jonas K Olofsson, Daniel A Broman, Paul E Gilbert, Pamela Dean, Steven Nordin, Claire Murphy
ABSTRACT
In experimental practice, odors are commonly applied to only one nostril for recordings of olfactory event-related potentials (OERPs), but the lateralization aspect of the OERP response is unclear regarding both stimulated nostril and cortical topography. The purpose of the present study was to investigate whether stimulated-nostril side affects OERP amplitudes and latencies and whether these potentials indicate lateralization of brain response in healthy, right-handed, young adults. OERPs were recorded from nine electrode sites in response to monorhinal stimulation with amyl acetate in 28 participants. The results showed a general increase in amplitude from frontal to parietal electrode sites (in particular for N1/P3) and generally larger amplitudes on the left hemisphere and midline than on the right hemisphere. There was no main effect of stimulated-nostril side on amplitude. Interactions indicated that N1/P2 amplitude was larger for left- than right-nostril stimulation and larger on the left hemisphere and midline than on the right hemisphere in left-nostril stimulation. No main effect or interactions of stimulated-nostril side over latencies were found and no effects on latencies of sagittal or coronal sites. These findings suggest a general parietal, left-hemisphere predominance in response amplitude to odorous stimulation and imply that either the left or the right nostril may be sufficient for accurate assessment of OERP latency in right-handed, young adults.
Sigma-Aldrich
Amyl acetate, ≥99%, FG
Sigma-Aldrich
Pentyl acetate, 99%
Sigma-Aldrich
Pentyl acetate, puriss. p.a., ≥98.5% (GC)