Merck
  • Sensitive high-performance liquid chromatographic assay for D-amino-acid oxidase activity in mammalian tissues using a fluorescent non-natural substrate, 5-fluoro-D-tryptophan.
Sensitive high-performance liquid chromatographic assay for D-amino-acid oxidase activity in mammalian tissues using a fluorescent non-natural substrate, 5-fluoro-D-tryptophan.
Journal of chromatography. A (2006-01-31)
Kenji Hamase, Reiko Nagayasu, Akiko Morikawa, Ryuichi Konno, Kiyoshi Zaitsu
ABSTRACT
A sensitive assay for D-amino-acid oxidase (DAO) activity in mammalian tissues has been established. D-Tryptophan (D-Trp) analogs were tested as substrates for DAO, and 5-fluoro-D-tryptophan (D-FTP) was found to be the best substrate. By the enzymatic reaction, D-FTP was converted to 5-fluoroindole-3-acetic acid (FIAA), a highly fluorescent product, and the product was determined by an RP-HPLC system with a fluorescence detector. The detection limit for purified DAO (from hog kidney) was 0.25 microU, and the within-day and day-to-day precisions of the assays were 4.6% (RSD, n=5), and 13.8% (RSD, 5 days), respectively. By the present method, the detailed distribution of DAO activity in the mouse brain was determined using individual animals for the first time, and significant activities were observed in the cerebellum, medulla oblongata and midbrain. Because sensitive DAO assay is frequently required in small tissues or in limited-tissue regions, the present method is useful for various research studies concerning DAO and the related D-amino acids.
Sigma-Aldrich
5-Fluoro-L-tryptophan, ≥98.0% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
5-Fluoro-DL-tryptophan, powder or crystals