Merck
  • Development of indole-3-propionic acid (OXIGON) for Alzheimer's disease.
Development of indole-3-propionic acid (OXIGON) for Alzheimer's disease.
Journal of molecular neuroscience : MN (2002-09-06)
Paul E Bendheim, Burkhard Poeggeler, Eyal Neria, Vivi Ziv, Miguel A Pappolla, Daniel G Chain
ABSTRACT
The accumulation of amyloid-beta and concomitant oxidative stress are major pathogenic events in Alzheimer's disease. Indole-3-propionic acid (IPA, OXIGON) is a potent anti-oxidant devoid of pro-oxidant activity. IPA has been demonstrated to be an inhibitor of beta-amyloid fibril formation and to be a potent neuroprotectant against a variety of oxidotoxins. This review will summarize the known properties of IPA and outline the rationale behind its selection as a potential disease-modifying therapy for Alzheimer's disease.
Sigma-Aldrich
3-Indolepropionic acid, ReagentPlus®, 99%
Sigma-Aldrich
Indole-3-propionic acid, ≥97.0% (T)
Sigma-Aldrich
Indole-3-propionic acid, ≥99.0% (T)