Merck
  • Studies on Lewis acid-mediated intramolecular cyclization reactions of allene-ene systems.
Studies on Lewis acid-mediated intramolecular cyclization reactions of allene-ene systems.
Chemical & pharmaceutical bulletin (2000-03-22)
K Hiroi, T Watanabe, A Tsukui
ABSTRACT
The Lewis acid-mediated reactions of allene-ene compounds, derived from 3-methylcitronellal or dimethyl malonate, were carried out using various Lewis acids such as ethylaluminum dichloride, diethylaluminum chloride, titanium chloride, zinc chloride etherate, or boron trifluoride etherate, affording unexpectedly intramolecular [2+2]cycloaddition products under some particular reaction conditions without any formation of intramolecular ene reaction products.
Sigma-Aldrich
Dimethyl malonate, 98%
Sigma-Aldrich
Dimethyl malonate, purum, ≥96.0% (GC)