Merck
  • Flow cytometric determination of Src phosphorylation in pediatric patients treated with dasatinib.
Flow cytometric determination of Src phosphorylation in pediatric patients treated with dasatinib.
Pediatric blood & cancer (2009-06-02)
Bella S Guerrouahen, Eric Wieder, Elisabeth G Blanchard, Francis Y Lee, Richard Aplenc, Seth J Corey
ABSTRACT
Tyrosine kinase inhibitors, such as imatinib, have dramatically improved the outcomes for patients with selected cancers. For imatinib, western blotting of phospho-CrkL was an insensitive, indirect, and descriptive method to determine drug efficacy. Greater use of targeted therapies should involve more quantitative evaluation of the target's dose-inhibition. The Src/Abl kinase inhibitor dasatinib has recently been approved for use in Ph+ leukemias after failure with imatinib. Src family kinases (SFK) also play a critical role in nonhematologic cancers. We have developed a flow cytometric assay to measure SFK autophosphorylation levels in blood mononuclear cells and observed a direct correlation between its inhibition and patient dosage. This method provides a sensitive, quick, and quantitative tool to assess drug efficacy.
Sigma-Aldrich
Histopaque®-1083, sterile-filtered, density: 1.083