Merck
  • Nudt21-mediated alternative polyadenylation of MZT1 3'UTR contributes to pancreatic cancer progression.
Nudt21-mediated alternative polyadenylation of MZT1 3'UTR contributes to pancreatic cancer progression.
iScience (2024-02-02)
Yu Zhou, Jiabin Yang, Leyi Huang, Chao Liu, Min Yu, Rufu Chen, Quanbo Zhou
ABSTRACT
Alternative polyadenylation (APA) is an important post-transcriptional regulatory mechanism and is involved in many diseases, but its function and mechanism in regulating pancreatic cancer (PC) pathogenesis remain unclear. In this study, we found that the 3' UTR shortening of MZT1 was the most prominent APA event in PC liver metastases. The short-3'UTR isoform exerted a stronger effect in promoting cell proliferation and migration both in vitro and in vivo. NUDT21, a core cleavage factor involved in APA, promoted the usage of proximal polyadenylation sites (PASs) on MZT1 mRNA by binding to the UGUA element located upstream of the proximal PAS. High percentage of distal polyA site usage index of MZT1 was significantly associated with a better prognosis. These findings demonstrate a crucial mechanism that NUDT21-mediated APA of MZT1 could promote the progression of PC. Our findings provided a better understanding of the connection between PC progression and APA machinery.
Sigma-Aldrich
Anti-MZT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution