Merck
  • Molecular mechanism of the interaction between Megalocytivirus-induced virus-mock basement membrane (VMBM) and lymphatic endothelial cells.
Molecular mechanism of the interaction between Megalocytivirus-induced virus-mock basement membrane (VMBM) and lymphatic endothelial cells.
Journal of virology (2023-10-25)
Jian-Hui He, Wenjie Shen, Deyu Han, Muting Yan, Mengting Luo, Hengwei Deng, Shaoping Weng, Jianguo He, Xiaopeng Xu
ABSTRACT
Viruses are able to mimic the physiological or pathological mechanism of the host to favor their infection and replication. Virus-mock basement membrane (VMBM) is a Megalocytivirus-induced extracellular structure formed on the surface of infected cells and structurally and functionally mimics the basement membrane of the host. VMBM provides specific support for lymphatic endothelial cells (LECs) rather than blood endothelial cells to adhere to the surface of infected cells, which constitutes a unique phenomenon of Megalocytivirus infection. Here, the structure of VMBM and the interactions between VMBM components and LECs have been analyzed at the molecular level. The regulatory effect of VMBM components on the proliferation and migration of LECs has also been explored. This study helps to understand the mechanism of LEC-specific attachment to VMBM and to address the issue of where the LECs come from in the context of Megalocytivirus infection.
Sigma-Aldrich
Anti-Phosphotyrosine antibody, Rabbit monoclonal, recombinant, expressed in HEK 293 cells, clone RM111, purified immunoglobulin
Sigma-Aldrich
Anti-GFP antibody produced in rabbit, affinity isolated antibody