Merck
  • SERPINB3-MYC axis induces the basal-like/squamous subtype and enhances disease progression in pancreatic cancer.
SERPINB3-MYC axis induces the basal-like/squamous subtype and enhances disease progression in pancreatic cancer.
Cell reports (2023-11-19)
Yuuki Ohara, Wei Tang, Huaitian Liu, Shouhui Yang, Tiffany H Dorsey, Helen Cawley, Paloma Moreno, Raj Chari, Mary R Guest, Azadeh Azizian, Jochen Gaedcke, Michael Ghadimi, Nader Hanna, Stefan Ambs, S Perwez Hussain
ABSTRACT
Pancreatic ductal adenocarcinoma (PDAC) exhibits distinct molecular subtypes: classical/progenitor and basal-like/squamous. Our study aimed to identify genes contributing to the development of the basal-like/squamous subtype, known for its aggressiveness. Transcriptome analyses revealed consistent upregulation of SERPINB3 in basal-like/squamous PDAC, correlating with reduced patient survival. SERPINB3 transgene expression in PDAC cells enhanced in vitro invasion and promoted lung metastasis in a mouse PDAC xenograft model. Metabolome analyses unveiled a metabolic signature linked to both SERPINB3 and the basal-like/squamous subtype, characterized by heightened carnitine/acylcarnitine and amino acid metabolism, associated with poor prognosis in patients with PDAC and elevated cellular invasiveness. Further analysis uncovered that SERPINB3 inhibited the cysteine protease calpain, a key enzyme in the MYC degradation pathway, and drove basal-like/squamous subtype and associated metabolic reprogramming through MYC activation. Our findings indicate that the SERPINB3-MYC axis induces the basal-like/squamous subtype, proposing SERPINB3 as a potential diagnostic and therapeutic target for this variant.
Sigma-Aldrich
Gemcitabine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-c-Myc antibody, Mouse monoclonal, clone 9E10, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-BBOX1 antibody produced in mouse, clone 6H3, purified immunoglobulin, buffered aqueous solution