Merck
  • Detection of IgA and IgG Antibodies against the Structural Proteins of SARS-CoV-2 in Breast Milk and Serum Samples Derived from Breastfeeding Mothers.
Detection of IgA and IgG Antibodies against the Structural Proteins of SARS-CoV-2 in Breast Milk and Serum Samples Derived from Breastfeeding Mothers.
Viruses (2023-04-28)
Karen Cortés-Sarabia, Vianey Guzman-Silva, Karla Montserrat Martinez-Pacheco, Jesús Alberto Meza-Hernández, Víctor Manuel Luna-Pineda, Marco Antonio Leyva-Vázquez, Amalia Vences-Velázquez, Fredy Omar Beltrán-Anaya, Oscar Del Moral-Hernández, Berenice Illades-Aguiar
ABSTRACT
Background: COVID-19 vaccination or natural infection is associated with the development of immunity. The search of IgA and IgG antibodies against all the structural proteins (spike, nucleocapsid, membrane, and envelope) of SARS-CoV-2 in breastfeeding mothers is associated with immunity that can help the newborn avoid development of the infection. Methods: In this study, we analyzed 30 breastfeeding women that provided samples of breast milk and serum and evaluated the presence of IgA, total IgG, and subclasses against the structural proteins of SARS-CoV-2. Results: We reported a high seroprevalence to IgA (76.67-100%) and negativity to IgG against all analyzed proteins in breast milk. Seroprevalence in serum samples was around 10-36.67% to IgA and 23.3-60% to IgG. Finally, we detected the presence of the subclasses IgG1, IgG2, and IgG4 against all the structural proteins of SARS-CoV-2. Conclusions: This work provides evidence of the presence of IgA and IgG antibodies against the four structural proteins of SARS-CoV-2 in breast milk and serum samples derived from breastfeeding women, which can confer immunity to the newborn.
Sigma-Aldrich
Anti-Human IgG1−Biotin antibody, Mouse monoclonal, clone 8c/6-39, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG3−Biotin antibody, Mouse monoclonal, clone HP-6050, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG2−Biotin antibody, Mouse monoclonal, clone HP-6014, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Human IgG4−Biotin antibody, Mouse monoclonal, clone HP-6025, purified from hybridoma cell culture