Merck
  • IL18 signaling causes islet β cell development and insulin secretion via different receptors on acinar and β cells.
IL18 signaling causes islet β cell development and insulin secretion via different receptors on acinar and β cells.
Developmental cell (2022-06-09)
Xian Zhang, Songyuan Luo, Minjie Wang, Qin Huang, Wenqian Fang, Jie Li, Tianxiao Liu, Yuanyuan Zhang, Zhiyong Deng, Cong-Lin Liu, Shuling Guan, Julio E Ayala, Richard A Flavell, Rohit N Kulkarni, Peter Libby, Junli Guo, Zhangsuo Liu, Guo-Ping Shi
ABSTRACT
Diabetic patients show elevated plasma IL18 concentrations. IL18 has two receptors: the IL18 receptor (IL18r) and the Na-Cl co-transporter (NCC). Here, we report that IL18 is expressed on islet α cells, NCC on β cells, and IL18r on acinar cells in human and mouse pancreases. The deficiency of these receptors reduces islet size, β cell proliferation, and insulin secretion but increases β cell apoptosis and exocrine macrophage accumulation after diet-induced glucose intolerance or streptozotocin-induced hyperglycemia. Together with the glucagon-like peptide-1 (GLP1), IL18 uses the NCC and GLP1 receptors on β cells to trigger β cell development and insulin secretion. IL18 also uses the IL18r on acinar cells to block hyperglycemic pancreas macrophage expansion. The β cell-selective depletion of the NCC or acinar-cell-selective IL18r depletion reduces glucose tolerance and insulin sensitivity with impaired β cell proliferation, enhanced β cell apoptosis and macrophage expansion, and inflammation in mouse hyperglycemic pancreas. IL18 uses NCC, GLP1r, and IL18r to maintain islet β cell function and homeostasis.
Sigma-Aldrich
EGF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Monoclonal Anti-α-Amylase 1A antibody produced in mouse, clone AMY-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-IL18R1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Thiazide-Sensitive NaCl Cotransporter Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Tamoxifen, ≥99%