Merck
  • An Optimally Fabricated Platform Guides Cancer-Specific Activation of Chemotherapeutic Drugs and Toxicity-Free Cancer Treatment.
An Optimally Fabricated Platform Guides Cancer-Specific Activation of Chemotherapeutic Drugs and Toxicity-Free Cancer Treatment.
Advanced healthcare materials (2022-06-08)
Ok Jeong Moon, Chul Joo Yoon, Bo-Ram Lee, Jeewon Lee
ABSTRACT
Cancer chemotherapeutic drugs such as doxorubicin, mitomycin C, and gemcitabine, which are mostly small synthetic molecules, are still clinically useful for cancer treatment. However, despite considerable therapeutic efficacy, severe toxicity-associated problems, which are mainly caused by the non-specific mode of action such as chromosomal DNA damage and interference in the DNA replication even in normal cells, remain unresolved and a major challenge for safer and thus more widespread adoption of chemotherapy. Herein, an innovative platform is developed through beneficially integrating core peptide units into highly-ordered, stable, and flexibly guest-adaptable structure of apoferritin, which simultaneously fulfills high-capacity loading of chemotherapeutic drugs compared with the case of FDA-approved antibody-drug conjugates, efficient drug targeting to cancer cells, and cancer cell-specific drug release and activation. This approach dramatically reduces drug toxicity to normal cells, significantly enhances efficacy in in vivo cancer treatment without toxicity to normal organs of mice, and thus is expected to open up a novel clinical route to break through the limits of current cancer chemotherapy.
Sigma-Aldrich
Gemcitabine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Cathepsin D from human liver, lyophilized powder, ≥250 units/mg protein (E1%/280)
Sigma-Aldrich
PrCP active human, recombinant, expressed in FreeStyle 293-F cells, ≥15% (SDS-PAGE)
Sigma-Aldrich
Cathepsin B Active human, recombinant, expressed in FreeStyle 293-F cells, ≥90% (SDS-PAGE)
Sigma-Aldrich
Cathepsin L Active human, recombinant, expressed in FreeStyle 293-F cells, ≥90% (SDS-PAGE)