Merck
  • Effect of angiotensin II and angiotensin-(1-7) on proliferation of stem cells from human dental apical papilla.
Effect of angiotensin II and angiotensin-(1-7) on proliferation of stem cells from human dental apical papilla.
Journal of cellular physiology (2020-06-11)
Larissa M Macedo, Renato I de Ávila, Gustavo R Pedrino, Diego B Colugnati, Marize C Valadares, Eliana M Lima, Clayton L Borges, Gregory T Kitten, Elisandra Gava, Carlos H Castro
ABSTRACT
The effects of the renin-angiotensin system (RAS) on stem cells isolated from human dental apical papilla (SCAPs) are completely unknown. Therefore, the aim of this study was to identify RAS components expressed in SCAPs and the effects of angiotensin (Ang) II and Ang-(1-7) on cell proliferation. SCAPs were collected from third molar teeth of adolescents and maintained in cell culture. Messenger RNA expression and protein levels of angiotensin-converting enzyme (ACE), ACE2, and Mas, Ang II type I (AT1) and type II (AT2) receptors were detected in SCAPs. Treatment with either Ang II or Ang-(1-7) increased the proliferation of SCAPs. These effects were inhibited by PD123319, an AT2 antagonist. While Ang II augmented mTOR phosphorylation, Ang-(1-7) induced ERK1/2 phosphorylation. In conclusion, SCAPs produce the main RAS components and both Ang II and Ang-(1-7) treatments induced cell proliferation mediated by AT2 activation through different intracellular mechanisms.
Sigma-Aldrich
Anti-H-CAM Antibody, clone F10-44-2, clone F10-44-2, Chemicon®, from mouse
Sigma-Aldrich
Anti-CD14 Antibody, clone 2D-15C, clone 2D-15C, Chemicon®, from mouse
Sigma-Aldrich
Anti-MCAM Antibody, clone P1H12, clone P1H12, Chemicon®, from mouse
Sigma-Aldrich
Anti-Human B cells (CD19) Antibody, clone FMC63, clone FMC63, Chemicon®, from mouse