Merck
  • Glibenclamide Treatment in Traumatic Brain Injury: Operation Brain Trauma Therapy.
Glibenclamide Treatment in Traumatic Brain Injury: Operation Brain Trauma Therapy.
Journal of neurotrauma (2020-11-19)
Ruchira M Jha, Stefania Mondello, Helen M Bramlett, C Edward Dixon, Deborah A Shear, W Dalton Dietrich, Kevin K W Wang, Zhihui Yang, Ronald L Hayes, Samuel M Poloyac, Philip E Empey, Audrey D Lafrenaye, Hong Q Yan, Shaun W Carlson, John T Povlishock, Janice S Gilsdorf, Patrick M Kochanek
ABSTRACT
Glibenclamide (GLY) is the sixth drug tested by the Operation Brain Trauma Therapy (OBTT) consortium based on substantial pre-clinical evidence of benefit in traumatic brain injury (TBI). Adult Sprague-Dawley rats underwent fluid percussion injury (FPI; n = 45), controlled cortical impact (CCI; n = 30), or penetrating ballistic-like brain injury (PBBI; n = 36). Efficacy of GLY treatment (10-μg/kg intraperitoneal loading dose at 10 min post-injury, followed by a continuous 7-day subcutaneous infusion [0.2 μg/h]) on motor, cognitive, neuropathological, and biomarker outcomes was assessed across models. GLY improved motor outcome versus vehicle in FPI (cylinder task, p < 0.05) and CCI (beam balance, p < 0.05; beam walk, p < 0.05). In FPI, GLY did not benefit any other outcome, whereas in CCI, it reduced 21-day lesion volume versus vehicle (p < 0.05). On Morris water maze testing in CCI, GLY worsened performance on hidden platform latency testing versus sham (p < 0.05), but not versus TBI vehicle. In PBBI, GLY did not improve any outcome. Blood levels of glial fibrillary acidic protein and ubiquitin carboxyl terminal hydrolase-1 at 24 h did not show significant treatment-induced changes. In summary, GLY showed the greatest benefit in CCI, with positive effects on motor and neuropathological outcomes. GLY is the second-highest-scoring agent overall tested by OBTT and the only drug to reduce lesion volume after CCI. Our findings suggest that leveraging the use of a TBI model-based phenotype to guide treatment (i.e., GLY in contusion) might represent a strategic choice to accelerate drug development in clinical trials and, ultimately, achieve precision medicine in TBI.
Sigma-Aldrich
Glyburide, meets USP testing specifications