Merck
  • The isoquinoline PRL-295 increases the thermostability of Keap1 and disrupts its interaction with Nrf2.
The isoquinoline PRL-295 increases the thermostability of Keap1 and disrupts its interaction with Nrf2.
iScience (2022-01-18)
Sharadha Dayalan Naidu, Takafumi Suzuki, Dina Dikovskaya, Elena V Knatko, Maureen Higgins, Miu Sato, Miroslav Novak, José A Villegas, Terry W Moore, Masayuki Yamamoto, Albena T Dinkova-Kostova
ABSTRACT
Transcription factor Nrf2 and its negative regulator Keap1 orchestrate a cytoprotective response against oxidative, metabolic, and inflammatory stress. Keap1 is a drug target, with several small molecules in drug development. Here, we show that the isoquinoline PRL-295 increased Keap1 thermostability in lysates from cells expressing fluorescently tagged Keap1. The thermostability of endogenous Keap1 also increased in intact cells and murine liver following PRL-295 treatment. Fluorescence Lifetime Imaging-Förster Resonance Energy Transfer (FLIM-FRET) experiments in cells co-expressing sfGFP-Nrf2 and Keap1-mCherry further showed that PRL-295 prolonged the donor fluorescence lifetime, indicating disruption of the Keap1-Nrf2 protein complex. Orally administered PRL-295 to mice activated the Nrf2transcriptional target NAD(P)H:quinone oxidoreductase 1 (NQO1) in liver and decreased the levels of plasma alanine aminotransferase and aspartate aminotransferase upon acetaminophen-induced hepatic injury. Thus, PRL-295 engages the Keap1 protein target in cells and in vivo, disrupting its interaction with Nrf2, leading to activation of Nrf2-dependent transcription and hepatocellular protection.
Sigma-Aldrich
Anti-Keap1 Antibody, clone 144, clone 144, from rat
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid