Merck
  • Tigecycline MIC testing by broth dilution requires use of fresh medium or addition of the biocatalytic oxygen-reducing reagent oxyrase to standardize the test method.
Tigecycline MIC testing by broth dilution requires use of fresh medium or addition of the biocatalytic oxygen-reducing reagent oxyrase to standardize the test method.
Antimicrobial agents and chemotherapy (2005-08-30)
Patricia A Bradford, Peter J Petersen, Mairead Young, C Hal Jones, Mark Tischler, John O'Connell
ABSTRACT
Tigecycline is a broad-spectrum glycylcycline antibiotic with activity against not only susceptible gram-positive and gram-negative pathogens but also strains that are resistant to many other antibiotics. In the process of determining quality control (QC) limits for the American Type Culture Collection reference strains for tigecycline, a number of inconsistencies in MICs were encountered which appeared to be related to the age of the Mueller-Hinton broth (MHB) medium used in the MIC testing. The objective of this study was to determine the cause of the discrepant MIC results between fresh and aged MHB. The MICs of tigecycline were determined in MHB that was either prepared fresh (<12 h old), prepared and stored at 4 degrees C, stored at room temperature, stored anaerobically, or supplemented with the biocatalytic oxygen-reducing reagent Oxyrase. When tested in fresh media, tigecycline was 2 to 3 dilutions more active against the CLSI-recommended QC strains compared to aged media (MICs of 0.03 to 0.25 and 0.12 to 0.5 mug/ml, respectively). Media aged under anaerobic conditions prior to testing or supplemented with Oxyrase resulted in MICs similar to those obtained in fresh medium (MICs of 0.03 to 0.12 and 0.03 to 0.25 mug/ml, respectively). Time-kill kinetics demonstrated a >3 log(10) difference in viable growth when tigecycline was tested in fresh or Oxyrase-supplemented MHB compared to aged MHB. High-pressure liquid chromatography analysis revealed the accumulation of an early peak (oxidative by-product of tigecycline) to be 3.5% in fresh media and 25.1% in aged media after 24 h and that addition of Oxyrase prevented the accumulation of this oxidized by-product. These results suggested that the activity of tigecycline was affected by the amount of dissolved oxygen in the media. The use of fresh MHB or supplementation with Oxyrase resulted in a more standardized test method for performing MIC tests with tigecycline.
Sigma-Aldrich
Oxyrase ® for Broth