Merck
Structure analysis of the receptor binding of 2019-nCoV.
Biochemical and biophysical research communications (2020-02-23)
Yun Chen, Yao Guo, Yihang Pan, Zhizhuang Joe Zhao
ABSTRACT
2019-nCoV is a newly identified coronavirus with high similarity to SARS-CoV. We performed a structural analysis of the receptor binding domain (RBD) of spike glycoprotein responsible for entry of coronaviruses into host cells. The RBDs from the two viruses share 72% identity in amino acid sequences, and molecular simulation reveals highly similar ternary structures. However, 2019-nCoV has a distinct loop with flexible glycyl residues replacing rigid prolyl residues in SARS-CoV. Molecular modeling revealed that 2019-nCoV RBD has a stronger interaction with angiotensin converting enzyme 2 (ACE2). A unique phenylalanine F486 in the flexible loop likely plays a major role because its penetration into a deep hydrophobic pocket in ACE2. ACE2 is widely expressed with conserved primary structures throughout the animal kingdom from fish, amphibians, reptiles, birds, to mammals. Structural analysis suggests that ACE2 from these animals can potentially bind RBD of 2019-nCoV, making them all possible natural hosts for the virus. 2019-nCoV is thought to be transmitted through respiratory droplets. However, since ACE2 is predominantly expressed in intestines, testis, and kidney, fecal-oral and other routes of transmission are also possible. Finally, antibodies and small molecular inhibitors that can block the interaction of ACE2 with RBD should be developed to combat the virus.
Sigma-Aldrich
SARS-COV-2-Spike-RBD epitope (450-469), 95% (HPLC), lyophilized powder
Sigma-Aldrich
Anti-SARS-COV-2-Spike-RBD region Peroxidase conjugated antibody produced in rabbit
Sigma-Aldrich
SARS-COV-2-Spike-RBD epitope (480-499), ≥95% (HPLC), lyophilized powder
Sigma-Aldrich
SARS-COV-2-Spike-RBD epitope (370-394), ≥95% (HPLC), lyophilized powder