Merck
  • Nickel ions attenuate autophagy flux and induce transglutaminase 2 (TG2) mediated post-translational modification of SQSTM1/p62.
Nickel ions attenuate autophagy flux and induce transglutaminase 2 (TG2) mediated post-translational modification of SQSTM1/p62.
Biochemical and biophysical research communications (2021-01-23)
Emi Aonuma, Akiko Tamura, Hiroki Matsuda, Takehito Asakawa, Yuriko Sakamaki, Kana Otsubo, Yoichi Nibe, Michio Onizawa, Yasuhiro Nemoto, Takashi Nagaishi, Kiichiro Tsuchiya, Tetsuya Nakamura, Motohiro Uo, Mamoru Watanabe, Ryuichi Okamoto, Shigeru Oshima
ABSTRACT
Nickel, the most frequent contact allergy cause, is widely used for various metallic materials and medical devices. Autophagy is an intracellular protein degradation system and contributes to metal recycling. However, it is unclear the functions of nickel in autophagy. We here demonstrated that NiCl2 induced microtubule-associated protein 1 light chain 3 (LC3)-II and LC3 puncta, markers of autophagosomes. Bafilomycin A1 (BafA1) treatment did not enhance LC3 puncta under NiCl2 stimulation, suggesting that NiCl2 did not induce autophagic flux. In addition, NiCl2 promotes the accumulation of SQSTM1/p62 and increased SQSTM1/p62 colocalization with lysosomal-associated membrane protein 1 (LAMP1). These data indicated that NiCl2 attenuates autophagic flux. Interestingly, NiCl2 induced the expression of the high-molecular-weight (MW) form of SQSTM1/p62. Inhibition of NiCl2-induced reactive oxygen species (ROS) reduced the high-MW SQSTM1/p62. We also showed that NiCl2-induced ROS activate transglutaminase (TG) activity. We found that transglutaminase 2 (TG2) inhibition reduced high-MW SQSTM1/p62 and SQSTM1/p62 puncta under NiCl2 stimulation, indicating that TG2 regulates SQSTM1/p62 protein homeostasis under NiCl2 stimulation. Our study demonstrated that nickel ion regulates autophagy flux and TG2 restricted nickel-dependent proteostasis.
Sigma-Aldrich
MISSION® esiRNA, targeting human TGM2
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-amide trifluoroacetate salt, ≥98% (HPLC), lyophilized powder
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by phenol extraction
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium bicarbonate, without sodium pyruvate, liquid, sterile-filtered, suitable for cell culture