Merck
  • High throughput proteomic and metabolic profiling identified target correction of metabolic abnormalities as a novel therapeutic approach in head and neck paraganglioma.
High throughput proteomic and metabolic profiling identified target correction of metabolic abnormalities as a novel therapeutic approach in head and neck paraganglioma.
Translational oncology (2021-06-13)
Zhigang Wang, Hongsai Chen, Lu Xue, Weiwei He, Wenying Shu, Hao Wu, Zhaoyan Wang
ABSTRACT
Head and neck paragangliomas (HNPGLs) are rare neoplasms that represent difficult treatment paradigms in neurotology. Germline mutations in genes encoding succinate dehydrogenase (SDH) are the cause of nearly all familial HNPGLs. However, the molecular mechanisms underlying tumorigenesis remain unclear. Mutational analysis identified 6 out of 14 HNPGLs harboring clinicopathologic SDH gene mutations. The SDHB gene was most frequently mutated in these patients, and western blot showed loss of SDHB protein in tumors with SDHB mutations. The paraganglioma cell line (PGL-626) was established from a sample that harbored a missense SDHB mutation (c.649C > T). Spectrometric analysis using tandem mass tags identified 151 proteins significantly differentially expressed in HNPGLs compared with normal nerves. Bioinformatics analyses confirmed the high level of enrichment of oxidative phosphorylation and metabolism pathways in HNPGLs. The mitochondrial complex subunits NDUFA2, NDUFA10, and NDUFA4, showed the most significantly increased expression and were localized predominantly in the cytoplasm of PGL-626 cells. The mitochondrial complex I inhibitor metformin exerted dose-dependent inhibitory effects on PGL-626 cells via cooperative down-regulation of NDUFA2, 4, and 10, with a significant decrease in the levels of reactive oxygen species and mitochondrial membrane potential. Further metabolomic analysis of PGL-626 cells showed that metabolites involved in central carbon metabolism in cancer and sphingolipid signaling pathways, pantothenate and CoA biosynthesis, and tryptophan and carbon metabolism were significantly altered after metformin treatment. Thus, this study provides insights into the molecular mechanisms underlying HNPGL tumorigenesis and identifies target correction of metabolic abnormalities as a novel therapeutic approach for this disease.
Sigma-Aldrich
Monoclonal Anti-Cyclin D1 antibody produced in mouse, clone DCS-6, ascites fluid
Sigma-Aldrich
Anti-SDHD antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-GAPDH antibody produced in mouse, purified immunoglobulin, buffered aqueous solution