Merck
  • TFG binds LC3C to regulate ULK1 localization and autophagosome formation.
TFG binds LC3C to regulate ULK1 localization and autophagosome formation.
The EMBO journal (2021-05-02)
Marianna Carinci, Beatrice Testa, Matteo Bordi, Giacomo Milletti, Massimo Bonora, Laura Antonucci, Caterina Ferraina, Marta Carro, Mukesh Kumar, Donatella Ceglie, Franziska Eck, Roberta Nardacci, Francois le Guerroué, Stefania Petrini, Maria E Soriano, Ignazio Caruana, Valentina Doria, Maria Manifava, Camille Peron, Matteo Lambrughi, Valeria Tiranti, Christian Behrends, Elena Papaleo, Paolo Pinton, Carlotta Giorgi, Nicholas T Ktistakis, Franco Locatelli, Francesca Nazio, Francesco Cecconi
ABSTRACT
The early secretory pathway and autophagy are two essential and evolutionarily conserved endomembrane processes that are finely interlinked. Although growing evidence suggests that intracellular trafficking is important for autophagosome biogenesis, the molecular regulatory network involved is still not fully defined. In this study, we demonstrate a crucial effect of the COPII vesicle-related protein TFG (Trk-fused gene) on ULK1 puncta number and localization during autophagy induction. This, in turn, affects formation of the isolation membrane, as well as the correct dynamics of association between LC3B and early ATG proteins, leading to the proper formation of both omegasomes and autophagosomes. Consistently, fibroblasts derived from a hereditary spastic paraparesis (HSP) patient carrying mutated TFG (R106C) show defects in both autophagy and ULK1 puncta accumulation. In addition, we demonstrate that TFG activity in autophagy depends on its interaction with the ATG8 protein LC3C through a canonical LIR motif, thereby favouring LC3C-ULK1 binding. Altogether, our results uncover a link between TFG and autophagy and identify TFG as a molecular scaffold linking the early secretion pathway to autophagy.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Anti-Atg13 Antibody, clone 2H4.2, clone 2H4.2, from mouse
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%