Merck
  • Synthesis of Low-Viscosity Ionic Liquids for Application in Dye-Sensitized Solar Cells.
Synthesis of Low-Viscosity Ionic Liquids for Application in Dye-Sensitized Solar Cells.
Chemistry, an Asian journal (2019-10-10)
Yanyan Fang, Pin Ma, Hongbo Cheng, Guoyu Tan, Jiaxin Wu, Jiaxin Zheng, Xiaowen Zhou, Shibi Fang, Yuhua Dai, Yuan Lin
ABSTRACT
Two types of ionic liquids (ILs), 1-(3-hexenyl)-3-methyl imidazolium iodide and 1-(3-butenyl)-3-methyl imidazolium iodide, are synthesized by introducing an unsaturated bond into the side alkyl chain of the imidazolium cation. These new ionic liquids exhibit high thermal stability and low viscosity (104 cP and 80 cP, respectively). The molecular dynamics simulation shows that the double bond introduced in the alkane chain greatly changes the molecular system space arrangement and diminishes the packing efficiency, leading to low viscosity. The low viscosity of the synthesized ionic liquids would enhance the diffusion of redox couples. This enhancement is detected by fabricating dye-sensitized solar cells (DSSCs) with electrolytes containing the two ILs and I2 . The highest efficiency of DSSCs is 6.85 % for 1-(3-hexenyl)-3-methyl imidazolium iodide and 5.93 % for 1-(3-butenyl)-3-methyl imidazolium iodide electrolyte, which is much higher than that of 5.17 % with the counterpart 1-hexyl-3-methyl imidazolium iodide electrolyte.
Sigma-Aldrich
1-Methylbenzimidazole, 99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
1 L
Please contact Customer Service for Availability
CA$82.40
To order products, please contact your local dealer.