Merck
  • Bmi1 Severs as a Potential Tumor-Initiating Cell Marker and Therapeutic Target in Esophageal Squamous Cell Carcinoma.
Bmi1 Severs as a Potential Tumor-Initiating Cell Marker and Therapeutic Target in Esophageal Squamous Cell Carcinoma.
Stem cells international (2020-09-05)
Xiaochen Wang, Kang Li, Maosheng Cheng, Ganping Wang, Hui Han, Fangfang Chen, Wenjing Liao, Zhi Chen, Jianwen Chen, Yong Bao, Liang Peng, Demeng Chen
ABSTRACT
Esophageal squamous cell carcinoma (ESCC) is a frequent malignant tumor with low 5-year overall survival. Targeting ESCC tumor-initiating cells (TICs) may provide a new research avenue to achieve better therapeutic effects of ESCC. However, the identity and characteristics of ESCC TICs remain poorly understood. Through genetic lineage tracing approach, we found that a group of Moloney murine leukemia virus insertion site 1- (Bmi1-) expressing cell populations present in the invasive front of the esophageal epithelium, providing a continuous flow of tumor cells for ESCC. Subsequently, we found that ablation of Bmi1+ cells from mice with ESCC led to inhibition of tumor growth. In addition, our results demonstrated that PTC-209, an inhibitor of Bmi1, was able to inhibit ESCC progression when combined with cisplatin. In summary, our data suggest that Bmi1+ cells serve as TICs in ESCC.
Sigma-Aldrich
Tamoxifen, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
4-Nitroquinoline N-oxide, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Cisplatin, British Pharmacopoeia (BP) Reference Standard