Merck
  • Natural Xanthone α-Mangostin Inhibits LPS-Induced Microglial Inflammatory Responses and Memory Impairment by Blocking the TAK1/NF-κB Signaling Pathway.
Natural Xanthone α-Mangostin Inhibits LPS-Induced Microglial Inflammatory Responses and Memory Impairment by Blocking the TAK1/NF-κB Signaling Pathway.
Molecular nutrition & food research (2020-06-09)
Huifeng Guan, Jiabing Li, Xiaofang Tan, Shenying Luo, Yangdan Liu, Yiwen Meng, Baichuan Wu, Yan Zhou, Yang Yang, Hongzhuan Chen, Lina Hou, Yu Qiu, Juan Li
ABSTRACT
The effect of α-mangostin (α-M), a polyphenolic xanthone isolated from mangostin, on lipopolysaccharide (LPS)-induced microglial activation and memory impairment is explored. The possible underlying mechanisms are also investigated. Cytokine production and activation of transforming growth factor activated kinase-1 (TAK1) and nuclear factor-κB (NF-κB) are detected by enzyme-linked immunosorbent assay (ELISA) or Western blot. Microglial migration and phagocytosis are evaluated with scratch wound-healing assay and phagocytosis of fluorescent latex beads, respectively. Learning and memory abilities of mice are evaluated with the Morris water maze test. The nanomolar (100-500 nm) α-M suppresses LPS-induced pro-inflammatory cytokine production and inducible nitric oxide synthase (iNOS) expression in microglia. It also inhibits LPS-induced microglial migration and phagocytosis. α-M rescues LPS-caused, microglia-mediated neuronal dendritic damage. Moreover, α-M represses LPS-induced toll-like receptor 4 (TLR4) expression and activation of TAK1 and NF-κB. In a mouse neuroinflammation model, α-M (50 mg kg-1 day-1 ) shows obvious anti-neuroinflammatory, neuroprotective, and memory-improving effects in vivo. α-M inhibits microglia-mediated neuroinflammation and prevents neurotoxicity and memory impairment from inflammatory damage. These results indicate that α-M has great potential to be used as a nutritional preventive strategy for neuroinflammation-related neurodegenerative disorders such as Alzheimer's disease.
Sigma-Aldrich
Latex beads, amine-modified polystyrene, fluorescent red, aqueous suspension, 1.0 μm mean particle size