Merck
  • Host cytosolic RNA sensing pathway promotes T Lymphocyte-mediated mycobacterial killing in macrophages.
Host cytosolic RNA sensing pathway promotes T Lymphocyte-mediated mycobacterial killing in macrophages.
PLoS pathogens (2020-05-29)
Yong Cheng, Nicholas J Kiene, Alexandra Tatarian, Emily F Eix, Jeffrey S Schorey
ABSTRACT
Mycobacterial infection leads to activation of the RIG-I/MAVS/TBK1 RNA sensing pathway in macrophages but the consequences of this activation remains poorly defined. In this study, we determined that activation of this RNA sensing pathway stimulates ICAM-1 expression in M.avium-infected macrophage through the inhibition of the E3 ubiquitin ligase CRL4COP1/DET1. CRL4 when active targets the transcription factor ETV5 for degradation by the ubiquitin-proteasome system. In the absence of the ETV5 transcription factor, ICAM-1 expression is significantly decreased. The M.avium-induced ICAM-1 production is required for the formation of immune synapse between infected macrophages and antigen-specific CD4+ T lymphocytes, and is essential for CD4+ T lymphocyte-mediated mycobacterial killing in vitro and in mice. This study demonstrates a previously undefined mechanism by which a host cytosolic RNA sensing pathway contributes to the interplay between mycobacteria infected macrophages and antigen-specific T lymphocytes.
Sigma-Aldrich
MISSION® esiRNA, targeting human IRF3
Sigma-Aldrich
MISSION® esiRNA, targeting human ETV5
Sigma-Aldrich
Fluorometric Intracellular Ros Kit, sufficient for 200 fluorometric tests (Deep red)
Sigma-Aldrich
MISSION® esiRNA, targeting human TBK1