Merck
  • Structural basis of lipid-driven conformational transitions in the KvAP voltage-sensing domain.
Structural basis of lipid-driven conformational transitions in the KvAP voltage-sensing domain.
Nature structural & molecular biology (2014-01-15)
Qufei Li, Sherry Wanderling, Pornthep Sompornpisut, Eduardo Perozo
ABSTRACT
Voltage-gated ion channels respond to transmembrane electric fields through reorientations of the positively charged S4 helix within the voltage-sensing domain (VSD). Despite a wealth of structural and functional data, the details of this conformational change remain controversial. Recent electrophysiological evidence showed that equilibrium between the resting ('down') and activated ('up') conformations of the KvAP VSD from Aeropyrum pernix can be biased through reconstitution in lipids with or without phosphate groups. We investigated the structural transition between these functional states, using site-directed spin-labeling and EPR spectroscopic methods. Solvent accessibility and interhelical distance determinations suggest that KvAP gates through S4 movements involving an ∼3-Å upward tilt and simultaneous ∼2-Å axial shift. This motion leads to large accessibly changes in the intracellular water-filled crevice and supports a new model of gating that combines structural rearrangements and electric-field remodeling.
Product Description
Avanti
16:0-10 Doxyl PC, Avanti Research - A Croda Brand 810603P, powder
Pricing and availability is not currently available.