Merck
  • Impact of artificial digestion on the sizes and shapes of microplastic particles.
Impact of artificial digestion on the sizes and shapes of microplastic particles.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2019-12-04)
Valerie Stock, Christoph Fahrenson, Andreas Thuenemann, Merve Hilal Dönmez, Linn Voss, Linda Böhmert, Albert Braeuning, Alfonso Lampen, Holger Sieg
ABSTRACT
Current analyses show a widespread occurrence of microplastic particles in food products and raise the question of potential risks to human health. Plastic particles are widely considered to be inert due to their low chemical reactivity and therefore supposed to pose, if at all only minor hazards. However, variable physicochemical conditions during the passage of the gastrointestinal tract gain strong importance, as they may affect particle characteristics. This study aims to analyze the impact of the gastrointestinal passage on the physicochemical particle characteristics of the five most produced and thus environmentally relevant plastic materials polyethylene, polypropylene, polyvinyl chloride, polyethylene terephthalate and polystyrene. Scanning electron microscopy (SEM) and subsequent image analysis were employed to characterize microplastic particles. Our results demonstrate a high resistance of all plastic particles to the artificial digestive juices. The present results underline that the main stages of the human gastrointestinal tract do not decompose the particles. This allows a direct correlation between the physicochemical particle characteristics before and after digestion. Special attention must be paid to the adsorption of organic compounds like proteins, mucins and lipids on plastic particles since it could lead to misinterpretations of particle sizes and shapes.
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Poly(vinyl chloride), high molecular weight
Sigma-Aldrich
Polypropylene, Isotactic, average Mw ~340,000, average Mn ~97,000