Merck
  • Cigarette smoke inhibits the NLRP3 inflammasome and leads to caspase-1 activation via the TLR4-TRIF-caspase-8 axis in human macrophages.
Cigarette smoke inhibits the NLRP3 inflammasome and leads to caspase-1 activation via the TLR4-TRIF-caspase-8 axis in human macrophages.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2020-01-10)
Marco Buscetta, Serena Di Vincenzo, Monica Miele, Ester Badami, Elisabetta Pace, Chiara Cipollina
ABSTRACT
The NLRP3 inflammasome is formed by the sensor NLRP3, the adaptor ASC, and pro-caspase-1. Assembly and activation of the inflammasome trigger caspase-1-dependent cleavage of pro-IL-1β and pro-IL-18 into their secreted forms. Cigarette smoke is a risk factor for chronic inflammatory diseases and is associated with macrophage dysfunction. The impact of cigarette smoke on NLRP3-dependent responses in macrophages is largely unknown. Herein, we investigated the effects of cigarette smoke extract (CSE) on the NLRP3 inflammasome in human monocyte-derived macrophages (MDMs) and THP-1 cells stimulated with lipopolysaccharide (LPS) and LPS plus the NLRP3 inflammasome activator ATP. We found that CSE inhibited the release of IL-1β and IL-18 as well as the expression of NLRP3 acting mainly at the transcriptional level. Interestingly, we found that CSE increased the caspase-1 activity via an NLRP3-independent and TLR4-TRIF-caspase-8-dependent pathway. Activation of caspase-1 by CSE led to a reduction of the basal glycolytic flux and impaired glycolytic burst in response to LPS. Overall, our findings unveil novel pathways leading to immune-metabolic alterations in human macrophages exposed to cigarette smoke. These mechanisms may contribute to macrophage dysfunction and increased risk of infection in smokers.
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by gel-filtration chromatography
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Caspase-8 Inhibitor II, The Caspase-8 Inhibitor II controls the biological activity of Caspase-8. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt hydrate, microbial, BioReagent, suitable for cell culture, ≥99% (HPLC)
Sigma-Aldrich
CP-456773 sodium salt, ≥98% (HPLC)