Merck
  • Genomic Aberrations that Activate D-type Cyclins Are Associated with Enhanced Sensitivity to the CDK4 and CDK6 Inhibitor Abemaciclib.
Genomic Aberrations that Activate D-type Cyclins Are Associated with Enhanced Sensitivity to the CDK4 and CDK6 Inhibitor Abemaciclib.
Cancer cell (2017-12-13)
Xueqian Gong, Lacey M Litchfield, Yue Webster, Li-Chun Chio, Swee Seong Wong, Trent R Stewart, Michele Dowless, Jack Dempsey, Yi Zeng, Raquel Torres, Karsten Boehnke, Cecilia Mur, Carlos Marugán, Carmen Baquero, Chunping Yu, Steven M Bray, Isabella H Wulur, Chen Bi, Shaoyou Chu, Hui-Rong Qian, Philip W Iversen, Farhana F Merzoug, Xiang S Ye, Christoph Reinhard, Alfonso De Dios, Jian Du, Charles W Caldwell, María José Lallena, Richard P Beckmann, Sean G Buchanan
ABSTRACT
Most cancers preserve functional retinoblastoma (Rb) and may, therefore, respond to inhibition of D-cyclin-dependent Rb kinases, CDK4 and CDK6. To date, CDK4/6 inhibitors have shown promising clinical activity in breast cancer and lymphomas, but it is not clear which additional Rb-positive cancers might benefit from these agents. No systematic survey to compare relative sensitivities across tumor types and define molecular determinants of response has been described. We report a subset of cancers highly sensitive to CDK4/6 inhibition and characterized by various genomic aberrations known to elevate D-cyclin levels and describe a recurrent CCND1 3'UTR mutation associated with increased expression in endometrial cancer. The results suggest multiple additional classes of cancer that may benefit from CDK4/6-inhibiting drugs such as abemaciclib.
Sigma-Aldrich
Anti-cdk2 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Retinoblastoma Protein (aa 773-928), Recombinant fusion protein containing the C-terminal fragment (residues 773-928) of human retinoblastoma protein (Rb) with an N-terminal His6-tag, expressed in E. coli.