Merck
Programmable RNA N6-methyladenosine editing by CRISPR-Cas9 conjugates.
Nature chemical biology (2019-08-07)
Xiao-Min Liu, Jun Zhou, Yuanhui Mao, Quanquan Ji, Shu-Bing Qian
ABSTRACT
RNA modification in the form of N6-methyladenosine (m6A) regulates nearly all the post-transcriptional processes. The asymmetric m6A deposition suggests that regional methylation may have distinct functional consequences. However, current RNA biology tools do not distinguish the contribution of individual m6A modifications. Here we report the development of 'm6A editing', a powerful approach that enables m6A installation and erasure from cellular RNAs without changing the primary sequence. We engineered fusions of CRISPR-Cas9 and a single-chain m6A methyltransferase that can be programmed with a guide RNA. The resultant m6A 'writers' allow functional comparison of single site methylation in different messenger RNA regions. We further engineered m6A 'erasers' by fusing CRISPR-Cas9 with ALKBH5 or FTO to achieve site-specific demethylation of RNAs. The development of programmable m6A editing not only expands the scope of RNA engineering, but also facilitates mechanistic understanding of epitranscriptome.
Sigma-Aldrich
Anti-Cas9 Antibody, clone 7A9, clone 7A9, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid